missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Zofenopril calcium, 98%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Quantity | 100 mg |
| Solubility | Solubility aq. soln.: .. Other solubilities: soluble in dmso, insoluble in methanol |
| Melting Point | 249.0°C to 251.0°C |
| Formula Weight | 448.58 |
| InChI Key | NSYUKKYYVFVMST-LETVYOFWSA-L |
| SMILES | [Ca++].C[C@H](CSC(=O)C1=CC=CC=C1)C(=O)N1C[C@H](C[C@H]1C([O-])=O)SC1=CC=CC=C1.C[C@H](CSC(=O)C1=CC=CC=C1)C(=O)N1C[C@H](C[C@H]1C([O-])=O)SC1=CC=CC=C1 |
| IUPAC Name | calcium bis((2S,4S)-1-[(2S)-3-(benzoylsulfanyl)-2-methylpropanoyl]-4-(phenylsulfanyl)pyrrolidine-2-carboxylate) |
| Molecular Weight (g/mol) | 897.16 |
| Chemical Name or Material | Zofenopril calcium |
Safety and Handling
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement Very toxic to aquatic life with long lasting effects. |
| Precautionary Statement | GHS P Statement Avoid release to the environment. Collect spillage. Dispose of contents/container to an approved waste disposal plant. |
RUO – Research Use Only