CAS RN 81938-43-4
IUPAC Name:
calcium bis((2S,4S)-1-[(2S)-3-(benzoylsulfanyl)-2-methylpropanoyl]-4-(phenylsulfanyl)pyrrolidine-2-carboxylate)
Synonyms:
calcium bis((2S,4S)-1-[(2S)-3-(benzoylsulfanyl)-2-methylpropanoyl]-4-(phenylsulfanyl)pyrrolidine-2-carboxylate)
Molecular Weight (g/mol):
897.16
Molecular Formula:
C44H44CaN2O8S4
InChi Key:
NSYUKKYYVFVMST-LETVYOFWSA-L
SMILES:
[Ca++].C[C@H](CSC(=O)C1=CC=CC=C1)C(=O)N1C[C@H](C[C@H]1C([O-])=O)SC1=CC=CC=C1.C[C@H](CSC(=O)C1=CC=CC=C1)C(=O)N1C[C@H](C[C@H]1C([O-])=O)SC1=CC=CC=C1
1
–
1
of
1
results
1
Thermo Scientific Chemicals Zofenopril calcium, 98%
CAS: 81938-43-4 Molecular Formula: 0·5 Ca