missing translation for 'onlineSavingsMsg'
Learn More

Zobell’s Solution, Reagents

Catalog No. 01333368
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01333368 1 L
01333367 4 L
01333366 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01333368 Supplier Labchem Inc Supplier No. CZ0001001B

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Zobell’s Solution, Reagent, use for Redox. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Zobell’s Solution
Packaging Opaque HDPE
CAS 7732-18-5, 7447-40-7, 14459-95-1
Specific Gravity 1.0048
Physical Form Liquid
Technique Redox
Color Yellow/Green
Boiling Point 100°C
For Use With (Equipment) Redox Meter
Synonym Oxidation Reduction Potential Standard
Solubility Completely miscible
Quantity 1 L
Grade Reagent
Show More Show Less