missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZNF483 Polyclonal Antibody

Description
Immunogen sequence: LLENLRNLEF LDFPVSKLEL ISQLKWVELP WLLEEVSKSS RLDESALDKI IERCLRDDDH GLMEESQQYC GSSEEDHGNQ GNSKG Highest antigen sequence identity to the following orthologs: Mouse - 41%, Rat - 40%.
ZNF483 may be involved in transcriptional regulation.
Spécifications
Spécifications
| Antigen | ZNF483 |
| Applications | Immunohistochemistry (Paraffin), Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | ZNF483 |
| Gene Accession No. | Q8TF39 |
| Gene Alias | KIAA1962; Zinc finger protein 483; zinc finger protein HIT-10; zinc finger protein with KRAB and SCAN domains 16; ZKSCAN16; ZNF483; ZSCAN48 |
| Gene Symbols | ZNF483 |
| Afficher plus de résultats |