missing translation for 'onlineSavingsMsg'
Learn More

Zirconocene Dichloride 97.0+%, TCI America™

Catalog No. Z00075G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
Z00075G 5 g
Z000725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. Z00075G Supplier TCI America Supplier No. Z00075G

Chemical Identifiers

CAS 1291-32-3
Molecular Formula C10H10Cl2Zr
MDL Number MFCD00003726
Synonym Bis(cyclopentadienyl)zirconium Dichloride

Specifications

Melting Point 244°C
Color White
Quantity 5 g
UN Number 3261
Formula Weight 292.31
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Zirconocene Dichloride
RTECSNumber ZH7525000
TSCA Yes