Learn More
Zirconium Standard For ICP, Assurance Grade, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Supplier: SPEX CertiPrep Group LLC PLZR22T
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
| Analyte | Concentration |
|---|---|
| Zr | 1000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
| Zirconium Standard | |
| Triple-leached LDPE bottle | |
| Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
| 2.0,0.25,97.75 | |
| Liquid | |
| AA, ICP | |
| Certified Reference Material | |
| Zr | |
| 2% HNO3 | |
| 233.247 | |
| 83761 | |
| 1000mg/L |
| For AA and ICP | |
| 13826-66-9 , 7697-37-2 , 7732-18-5 | |
| 0.1,2.0,97.9 | |
| Zr | |
| H2N2O7Zr | |
| AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
| UJVRJBAUJYZFIX-UHFFFAOYSA-N | |
| [N+](=O)(O)[O-].[N+](=O)(O)[O-].O=[Zr] | |
| nitric acid;oxozirconium | |
| 250 mL | |
| Assurance |
Chemical Identifiers
| 13826-66-9 | |
| 233.247 | |
| 83761 | |
| [N+](=O)(O)[O-].[N+](=O)(O)[O-].O=[Zr] |
| H2N2O7Zr | |
| UJVRJBAUJYZFIX-UHFFFAOYSA-N | |
| nitric acid;oxozirconium |
Safety and Handling
Danger: Causes severe skin burns and eye damage Do not breathe dusts or mists. IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower. If in eyes, rinse cautiously with water for several minutes. Remove
DOTInformation : Hazardous
Recommended Storage : Ambient
Product intended for laboratory use only