missing translation for 'onlineSavingsMsg'
Learn More

Zirconium Standard For ICP, Assurance Grade, SPEX CertiPrep™

Catalog No. PLZR22Y
Change view
Click to view available options
Quantity:
125 mL
250 mL
500 mL
Packaging:
Triple-leached LDPE bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
PLZR22Y 125 mL Triple-leached LDPE bottle
PLZR22T 250 mL Triple-leached LDPE bottle
PLZR22X 500 mL Triple-leached LDPE bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PLZR22Y Supplier SPEX CertiPrep Group LLC Supplier No. PLZR22Y

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Zr1000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Chemical Identifiers

CAS 13826-66-9 , 7697-37-2 , 7732-18-5
Molecular Formula H2N2O7Zr
Molecular Weight (g/mol) 233.247
InChI Key UJVRJBAUJYZFIX-UHFFFAOYSA-N
PubChem CID 83761
IUPAC Name nitric acid;oxozirconium
SMILES [N+](=O)(O)[O-].[N+](=O)(O)[O-].O=[Zr]

Specifications

Packaging Triple-leached LDPE bottle
Certificate of Analysis Assurance Certificate of Analysis reports up to 39 elements at ppb levels.
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
CAS Min % 0.1, 2.0, 97.9
CAS Max % 0.1, 2.0, 97.9
Physical Form Liquid
Technique AA, ICP
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Type Certified Reference Material
Solvent or Matrix 2% HNO3
Quantity 125 mL
Grade Assurance
Concentration or Composition (by Analyte or Components) 1000mg/L
Chemical Name or Material Zirconium Standard, For AA and ICP
Show More Show Less
Hazard Category Danger: Causes severe skin burns and eye damage Do not breathe dusts or mists. IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower. If in eyes, rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a poison center/doctor. Store locked up. Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Hazardous
Recommended Storage Ambient

Product intended for laboratory use only