missing translation for 'onlineSavingsMsg'
Learn More

Zirconium Standard For ICP, 2% HNO3 in 1000μg/mL, SPEX CertiPrep™

Catalog No. PLZR22M
Change view
Click to view available options
Quantity:
30 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PLZR22M 30 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PLZR22M Supplier SPEX CertiPrep Group LLC Supplier No. PLZR22M

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Zr1000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Chemical Identifiers

CAS 13826-66-9 , 7697-37-2 , 7732-18-5
Molecular Formula H2N2O7Zr
Molecular Weight (g/mol) 233.247
InChI Key UJVRJBAUJYZFIX-UHFFFAOYSA-N
PubChem CID 83761
IUPAC Name nitric acid;oxozirconium
SMILES [N+](=O)(O)[O-].[N+](=O)(O)[O-].O=[Zr]

Specifications

Packaging Triple-leached LDPE bottle
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
CAS Max % 2, 0.25, 97.75
Physical Form Liquid
Technique AA, ICP
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Type Certified Reference Material
Solvent or Matrix 2% HNO3
Quantity 30 mL
Grade Assurance
Concentration or Composition (by Analyte or Components) 1000mg/L
Chemical Name or Material Zirconium Standard, For AA and ICP
Show More Show Less
Hazard Category May cause cancer.
DOTInformation Nonhazardous
Recommended Storage Ambient

Product intended for laboratory use only