missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zirconium Standard For ICP, 10,000μg/mL in 5% HNO3, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Supplier: SPEX CertiPrep Group LLC PLZR23X
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
| Analyte | Concentration |
|---|---|
| Zr | 10,000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
| Zirconium Standard | |
| Assurance Certificate of Analysis reports up to 30 elements at ppb levels | |
| Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
| AA, ICP | |
| Certified Reference Material | |
| [N+](=O)(O)[O-].[N+](=O)(O)[O-].O=[Zr] | |
| nitric acid;oxozirconium | |
| 500 mL | |
| Assurance |
| Triple-leached LDPE bottle | |
| 13826-66-9 , 7697-37-2 , 7732-18-5 | |
| H2N2O7Zr | |
| AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
| UJVRJBAUJYZFIX-UHFFFAOYSA-N | |
| 5% HNO3 | |
| 233.247 | |
| 83761 |
Chemical Identifiers
| 13826-66-9 | |
| 233.247 | |
| 83761 | |
| [N+](=O)(O)[O-].[N+](=O)(O)[O-].O=[Zr] |
| H2N2O7Zr | |
| UJVRJBAUJYZFIX-UHFFFAOYSA-N | |
| nitric acid;oxozirconium |
Safety and Handling
Causes severe skin burns and eye damage
DOTInformation : Hazardous
Recommended Storage : Ambient
Product intended for laboratory use only