missing translation for 'onlineSavingsMsg'
Learn More

Zirconium sponge, 0.8-25.4mm (0.03-1.0in), 99.5%, Zr & Hf, Thermo Scientific Chemicals

Catalog No. AA10441A1
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA10441A1 1 kg
AA1044118 50 g
AA1044130 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA10441A1 Supplier Thermo Scientific Chemicals Supplier No. 010441A1

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-67-7
Molecular Formula Zr
Molecular Weight (g/mol) 91.22
MDL Number MFCD00011303 MFCD00148878
InChI Key QCWXUUIWCKQGHC-UHFFFAOYSA-N
Synonym zircat, powder, circonio, zirconium, elemental, zirconio, zirkonium, compounds, unii-c6v6s92n3c, and compounds, foil
PubChem CID 23995
ChEBI CHEBI:33342
IUPAC Name zirconium
SMILES [Zr]

Specifications

Color Gray
Appearance Gray sponge
Quantity 1 kg
Odor Odorless
Form Sponge
Assay Percent Range 99.5%
Chemical Name or Material Zirconium
Hazard Category H250-H260
Precautionary Statement P210-P222-P223-P231+P232-P280-P335+P334-P370+P378q-P501c
DOTInformation DOT Class: 4.2; Packaging Group: II
EINECSNumber 231-176-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only