missing translation for 'onlineSavingsMsg'
Learn More

Zirconium Oxide, Approx. 99%, Spectrum™ Chemical

Catalog No. 18614831
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
45 kg
Packaging:
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18614831 500 g Poly Pail
18614828 125 g Poly Pail
18614829 2.5 kg Poly Pail
18614827 12 kg Poly Pail
18614830 45 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18614831 Supplier Spectrum Chemical Mfg Cor Supplier No. Z1102500GM
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Reagent grade zirconium oxide powder with approximately 99% purity.

  • Grade: Reagent
  • Purity: ∼99%
  • Appearance: Powder Crystals
  • Odor: Odorless
  • Molecular Weight: 123.22
  • Molecular Formula: ZrO2
  • CAS Number: 1314-23-4
  • Melting Point: 2680°C to 2715°C
  • Boiling Point: 4300°C
  • Relative Density: 5.68 to 5.85
  • Chemical Stability: Stable under normal conditions
  • Incompatible materials: Strong oxidizing agents, strong acids, and strong bases
  • Solubility: Insoluble in water; slightly soluble in Nitric Acid
  • Chemical Name: Zirconium Oxide
  • Color: White
  • Physical State: Solid (Powder Crystals)
  • Density: 5.68 to 5.85

Chemical Identifiers

CAS 1314-23-4

Specifications

Boiling Point 4300°C
Melting Point 2680°C to 2715°C
Color White
CAS Max % 1
Physical Form Crystalline Powder or Needles
Quantity 500 g
Assay Percent Range 99%
Molecular Formula ZrO2
Linear Formula ZrO2
Synonym Zirconium Dioxide
Solubility Information Insoluble in water, slightly soluble in nitric acid
SMILES [O-2].[O-2].[Zr+4]
IUPAC Name zirconium;oxide
Molecular Weight (g/mol) 123.22
Percent Purity ∼99%
Grade Reagent
Manufacturing Origin USA
Odor Odorless
Packaging Poly Pail
Appearance Powder Crystals
Density 5.68 to 5.85 g/cm³
Chemical Name or Material Zirconium Oxide
Show More Show Less

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.