missing translation for 'onlineSavingsMsg'
Learn More

Zirconium(IV) Propoxide (ca. 70% in 1-Propanol), TCI America™

Catalog No. Z001725G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
Z001725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. Z001725G Supplier TCI America Supplier No. Z001725G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Flammable zirconium(IV) propoxide solution (approx. 70% in 1-propanol) used as a laboratory reagent.

  • Chemical Formula: C12H28O4Zr
  • Formula Weight: 327.58
  • Concentration: 70.0 to 75.0 w/w% in 1-Propanol
  • Physical State: Liquid
  • Flash Point: 28 °C
  • Storage Temperature: Recommended in a cool and dark place, <15°C; store under inert gas
  • Moisture sensitive

Chemical Identifiers

CAS 23519-77-9
Molecular Formula C12H28O4Zr
Molecular Weight (g/mol) 327.58
MDL Number MFCD00015307
InChI Key XPGAWFIWCWKDDL-UHFFFAOYSA-N
Synonym zirconium iv tetrapropoxide, zirconium iv propoxide, unii-a7z4n4e80x, 1-propanol, zirconium 4+ salt, zirconium propoxide, zirconium n-propoxide, 1-propanol, zirconium 4+ salt 4:1, zirconium iv propoxide solution, zirconium tetrapropanolate, propyl alcohol, zirconium 4+ salt
PubChem CID 90139
IUPAC Name tetrapropoxyzirconium
SMILES CCCO[Zr](OCCC)(OCCC)OCCC

Specifications

Color Yellow
CAS Min % 70.0, ≥30%
CAS Max % 75, < 40
Physical Form Liquid
Quantity 25 g
Linear Formula Zr(OCH2CH2CH3)4
UN Number 1274
Beilstein 3935210
Flash Point 28°C (82.4°F)
Solubility Information Solubility in Toluene, Alcohol
InChI Formula InChI=1S/C12H28O4Zr/c1-5-9-13-16(14-10-6-2,15-11-7-3)12-8-4/h5-12H2,1-4H3
Formula Weight 327.58
Concentration or Composition (by Analyte or Components) Zirconium(IV) Propoxide: 70.0 to 75.0 w/w% in 1-Propanol
Refractive Index 1.45
Sensitivity Moisture Sensitive
Appearance White to yellow to green clear liquid
Specific Gravity 1.04 (at 20/20°C)
Chemical Name or Material Zirconium(IV) Propoxide (ca. 70% in 1-Propanol)
Show More Show Less
EINECSNumber (2)-0226
TSCA Yes