missing translation for 'onlineSavingsMsg'
Learn More

Zirconium ICP Standard, 10,000 ppm Zr in 5% HNO3, Ricca Chemical

$1,138.43 - $1,138.43

Chemical Identifiers

CAS 13826-66-9
Molecular Formula H2N2O7Zr
Molecular Weight (g/mol) 233.247
InChI Key UJVRJBAUJYZFIX-UHFFFAOYSA-N
PubChem CID 83761
IUPAC Name nitric acid;oxozirconium
SMILES [N+](=O)(O)[O-].[N+](=O)(O)[O-].O=[Zr]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
PZR10KN-500
View Documents
Ricca Chemical Company
PZR10KN500
500 mL Natural Poly Bottle N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

13826-66-9
233.247
83761
[N+](=O)(O)[O-].[N+](=O)(O)[O-].O=[Zr]
H2N2O7Zr
UJVRJBAUJYZFIX-UHFFFAOYSA-N
nitric acid;oxozirconium

Specifications

Natural Poly Bottle
89.0,6.68,2.43
93.0,6.95,2.53
Liquid
Colorless
Standard
[N+](=O)(O)[O-].[N+](=O)(O)[O-].O=[Zr]
233.247
83761
13826-66-9 , 7697-37-2 , 7732-18-5
10,000ppm Zr
1 mL = 10 mg Zr
H2N2O7Zr
< 2
UJVRJBAUJYZFIX-UHFFFAOYSA-N
nitric acid;oxozirconium
500 mL
Zirconium ICP Standard
Videos
SDS
Documents

Documents