missing translation for 'onlineSavingsMsg'
Learn More

Zirconium ICP Standard, 10,000 ppm Zr in 5% HNO3, Ricca Chemical

Catalog No. PZR10KN500
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Natural Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
PZR10KN500 500 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PZR10KN500 Supplier Ricca Chemical Company Supplier No. PZR10KN500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 13826-66-9 , 7697-37-2 , 7732-18-5
Molecular Formula H2N2O7Zr
Molecular Weight (g/mol) 233.247
InChI Key UJVRJBAUJYZFIX-UHFFFAOYSA-N
PubChem CID 83761
IUPAC Name nitric acid;oxozirconium
SMILES [N+](=O)(O)[O-].[N+](=O)(O)[O-].O=[Zr]

Specifications

Packaging Natural Poly Bottle
Concentration or Composition Notes 10,000ppm Zr
CAS Min % 89.0, 6.68, 2.43
CAS Max % 93.0, 6.95, 2.53
Equivalence 1 mL = 10 mg Zr
Physical Form Liquid
Color Colorless
pH < 2
Type Standard
Quantity 500 mL
Chemical Name or Material Zirconium ICP Standard
Show More Show Less