Learn More
Abcam Inc Zinquin, Zn2+ selective fluorescent probe, 5MG
Numéro de catalogue. 502705180
Encompass
MW 386.4 Da, Purity >95%. Intracellular Zn2+ selective fluorescent probe. Forms zinquin-Zn2+ complexes in either 1:1 or 2:1 ratios (Kd values are 370 and 850 nM respectively). Zinquin ethyl ester (ab145478) analog. Labels micromolar concentrations of intracellular Zn2+ ions with an intense extranuclear fluorescence emitting in the blue range.
The product is subject to the following: Abcam Restricted Use Statement
Numéro de catalogue. 502705180 Fournisseur Abcam Inc Code fournisseur AB1454515MG
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
MW 386.4 Da, Purity >95%. Intracellular Zn2+ selective fluorescent probe. Forms zinquin-Zn2+ complexes in either 1:1 or 2:1 ratios (Kd values are 370 and 850 nM respectively). Zinquin ethyl ester (ab145478) analog. Labels micromolar concentrations of intracellular Zn2+ ions with an intense extranuclear fluorescence emitting in the blue range.
The product is subject to the following: Abcam Restricted Use Statement