missing translation for 'onlineSavingsMsg'
Learn More

Zinc tert-butoxide

Catalog No. AA40718MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA40718MD 250 mg
AA4071803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA40718MD Supplier Thermo Scientific Chemicals Supplier No. 040718MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4278-43-7 | C8H18O2Zn | 211.61 g/mol

A sol-gel process has been developed for the chemical synthesis of ZnS at room temperature using zinc tert-butoxide and H 2 S as precursors in a toluene solution. The key step that enables this zinc-mediated epoxidation to be catalytic may be the equilibrium between zinc peroxide (R)-15 and zinc tert-butoxide (R)-16 in the presence of t-BuOOH. An additional important feature that renders the dual catalytic cycles feasible is the poor reactivity of zinc tert-butoxide toward the iridium-bound cationic intermediate, whereby the requisite deprotonation process is effectively mediated by zinc tert-butoxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4278-43-7
Molecular Formula C8H18O2Zn
Molecular Weight (g/mol) 211.61
MDL Number MFCD00145545
InChI Key DVGVEVPHJQJMPP-UHFFFAOYSA-N
Synonym zinc tert-butoxide, di-tert-butoxyzinc, acmc-20aj87, zinc bis 2-methylpropan-2-olate, zinc 2+ bis 2-methylpropan-2-olate, zinc 2+ ion bis 2-methylpropan-2-olate
PubChem CID 14178434
IUPAC Name zinc;2-methylpropan-2-olate
SMILES CC(C)(C)[O-].CC(C)(C)[O-].[Zn+2]

Specifications

Color White to Yellow
Odor Odorless
Linear Formula Zn[OC(CH3)3]2
Quantity 250 mg
UN Number UN3181
Sensitivity Moisture sensitive
Solubility Information Insoluble in water.
Formula Weight 211.61
Physical Form Powder
Chemical Name or Material Zinc tert-butoxide
Hazard Category H228-H315-H319-H335
Hazard Statement GHS H Statement
H228-H315-H319-H335
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P240-P241-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL SALTS OF ORGANIC COMPOUNDS, FLAMMABLE, N.O.S.
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only