missing translation for 'onlineSavingsMsg'
Learn More

Zinc sulfate hydrate, Puratronic, 99.998% (metals basis)

Catalog No. AA1113809
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1113809 10 g
AA1113818 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1113809 Supplier Thermo Scientific Chemicals Supplier No. 01113809
Only null left

CAS: 16788-42-4 | H2O5SZn | 179.45 g/mol

Zinc sulfate hydrate is used in the production of rayon, as a feed supplement, and as a fertilizer ingredient.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 16788-42-4
Molecular Formula H2O5SZn
Molecular Weight (g/mol) 179.45
MDL Number MFCD00149893
InChI Key RNZCSKGULNFAMC-UHFFFAOYSA-L
Synonym zinc sulfate monohydrate, zinc sulfate hydrate, unii-ptx099xsf1, ptx099xsf1, sulfuric acid, zinc salt 1:1 , monohydrate, zinc sulphate monohydrate, gunningite, sulfato de zinc monohydratado, zinc sulfate tn, zinc sulfate,monohydrate
PubChem CID 62639
SMILES O.[Zn++].[O-]S([O-])(=O)=O

Specifications

Melting Point 100°C
Color White
Physical Form Crystalline Powder
Quantity 10 g
Linear Formula ZnSO4·xH2O
UN Number UN3077
Merck Index 14,10159
Solubility Information Soluble in water.
IUPAC Name zinc;sulfate;hydrate
Formula Weight 161.43 (Anhydrous)
Percent Purity 99.999%
Grade Puratronic
Odor Odorless
Assay (metals basis)
Chemical Name or Material Zinc sulfate hydrate
Show More Show Less
Hazard Category H302-H318
Hazard Statement GHS H Statement
H318-H302
Causes serious eye damage.
Harmful if swallowed.
Precautionary Statement P264b-P270-P280-P301+P312-P305+P351+P338-P310-P330-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 231-793-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only