missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc Sulfate, For Nitrogen (Ammonia), Nitrogen (Organic), 10% (w/v), LabChem™
Supplier: LabChem LC272302
Specifications
| Zinc Sulfate | |
| For Nitrogen (Ammonia), Nitrogen (Organic) | |
| 83.76,16.24 | |
| Zn(NO3)2·6H2O | |
| 1.1g/mL | |
| Soluble in water | |
| O.O.O.O.O.O.O.[Zn++].[O-]S([O-])(=O)=O | |
| 287.54 | |
| CHEBI:32312 | |
| 10% (w/v) | |
| Poly Bottle | |
| 1.35cSt | |
| 1 L |
| Colorless | |
| 7446-20-0,7732-18-5 | |
| H14O11SZn | |
| MFCD00149894 | |
| zinc sulfate heptahydrate, zinc sulfate jan, zinc sulfate 1:1 heptahydrate, unii-n57ji2k7wp, zinc vitriol heptahydrate, zinc sulfate heptahydrate 1:1:7, white vitriol heptahydrate, ccris 5563, zinc sulfate znso4 heptahydrate, znso4.7h2o | |
| RZLVQBNCHSJZPX-UHFFFAOYSA-L | |
| zinc(2+) heptahydrate sulfate | |
| 62640 | |
| 297.48 | |
| Passes Test | |
| 1.1g/mL | |
| Sulfur compounds | |
| Liquid |
Chemical Identifiers
| 7446-20-0 | |
| 287.54 | |
| RZLVQBNCHSJZPX-UHFFFAOYSA-L | |
| 62640 | |
| zinc(2+) heptahydrate sulfate |
| H14O11SZn | |
| MFCD00149894 | |
| zinc sulfate heptahydrate, zinc sulfate jan, zinc sulfate 1:1 heptahydrate, unii-n57ji2k7wp, zinc vitriol heptahydrate, zinc sulfate heptahydrate 1:1:7, white vitriol heptahydrate, ccris 5563, zinc sulfate znso4 heptahydrate, znso4.7h2o | |
| CHEBI:32312 | |
| O.O.O.O.O.O.O.[Zn++].[O-]S([O-])(=O)=O |
Safety and Handling
GHS H Statement
Toxic to aquatic life.
GHS P Statement
Avoid release to the environment.
Dispose of contents/container to comply with local, state and federal regulations.
Recommended Storage : Room Temperature