missing translation for 'onlineSavingsMsg'
Learn More

Zinc stearate

Catalog No. AC263780200
Change view
Click to view available options
Quantity:
1 kg
5 kg
20 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC263780200 20 kg
AC263780010 1 kg
AC263780050 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC263780200 Supplier Thermo Scientific Chemicals Supplier No. 263780200

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 557-05-1 | C36H70O4Zn | 632.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 557-05-1
MDL Number MFCD00013031
InChI Key XOOUIPVCVHRTMJ-UHFFFAOYSA-L
Synonym zinc stearate, zinc distearate, zinc octadecanoate, stearates, hydense, metallac, hytech, mathe, coad, zink distearat
PubChem CID 11178
IUPAC Name zinc;octadecanoate
SMILES CCCCCCCCCCCCCCCCCC(=O)[O-].CCCCCCCCCCCCCCCCCC(=O)[O-].[Zn+2]

Specifications

Melting Point 118.0°C to 128.0°C
Color White
Flash Point >100°C
Infrared Spectrum Authentic
Packaging Fibre drum
Molecular Formula C36H70O4Zn
Linear Formula [CH3(CH2)16CO2]2Zn
Quantity 20 kg
Merck Index 15, 10358
Solubility Information Solubility in water: insoluble. Other solubilities: insoluble in ethanol,diethyl ether,,soluble in benzene,xylene,fats,oils
Molecular Weight (g/mol) 632.34
Formula Weight 632.34
Percent Purity 12.5 to 14.0% (ZnO)
Physical Form Powder
Chemical Name or Material Zinc stearate
Show More Show Less
EINECSNumber 209-151-9
RTECSNumber ZH5200000
TSCA TSCA

RUO – Research Use Only