Learn More
Merck Emd Millipore Zinc plated pump, each
Numéro de catalogue. NC0821369
Encompass
This Rieke pump is for dispensing solvents and non-corrosive liquids. The self-venting design eliminates a need for an air vent opening. Pump includes a plastic Flex-spout mounting cap and a stabilizing metal strap for mounting to Sigma-Aldrich 18L-CS or 20 L Flex-spout cans. Pump may also be used with Sigma-Aldrich 18L-CS or 20 L NPT threaded cans with 2 in. bung (NPT adapter Z557811 is required).Dispenses 100 mL/stroke.
Numéro de catalogue. NC0821369 Fournisseur Merck Emd Millipore Code fournisseur Z5578031EA
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
This Rieke pump is for dispensing solvents and non-corrosive liquids. The self-venting design eliminates a need for an air vent opening. Pump includes a plastic Flex-spout mounting cap and a stabilizing metal strap for mounting to Sigma-Aldrich 18L-CS or 20 L Flex-spout cans. Pump may also be used with Sigma-Aldrich 18L-CS or 20 L NPT threaded cans with 2 in. bung (NPT adapter Z557811 is required).Dispenses 100 mL/stroke.