missing translation for 'onlineSavingsMsg'
Learn More

Zinc Metal, 0.25 in. and Finer, Shot, 99.8%, Spectrum™ Chemical

Catalog No. 18614750
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
Packaging:
Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18614750 12 kg Poly Bottle
18614748 500 g Poly Bottle
18614749 2.5 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18614750 Supplier Spectrum Chemical Mfg Cor Supplier No. Z103512KG
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7440-66-6
Molecular Weight (g/mol) 65.39

Specifications

CAS Max % 1
Quantity 12 kg
Packaging Poly Bottle
Particle Size ≤0.25 in.
Form Shot
Percent Purity 99.8%
Grade Reagent
Chemical Name or Material Zinc Metal

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.