missing translation for 'onlineSavingsMsg'
Learn More

Zinc, Granular, 20 Mesh, ACS Reagent Grade, Ricca Chemical

Catalog No. RDCZ0050100
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
White Poly Wide Mouth Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
RDCZ0050100 100 g White Poly Wide Mouth Bottle
RDCZ0050500 500 g White Poly Wide Mouth Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. RDCZ0050100 Supplier Ricca Chemical Company Supplier No. RDCZ0050100B1

Chemical Identifiers

CAS 7440-66-6
Molecular Formula Zn
Molecular Weight (g/mol) 65.38
MDL Number MFCD00011291
InChI Key HCHKCACWOHOZIP-UHFFFAOYSA-N
Synonym dust, cinc, powder, merrillite, rheinzink, jasad, blue powder, granular, zinc, elemental, emanay dust
PubChem CID 23994
ChEBI CHEBI:30185
IUPAC Name zinc
SMILES [Zn]

Specifications

CAS Min % 98.0
CAS Max % 100.0
Quantity 100 g
Packaging White Poly Wide Mouth Bottle
Form Fine Granular Powder
Percent Purity Min. 99.8%
Grade ACS
Concentration or Composition (by Analyte or Components) 20 Mesh
Chemical Name or Material Zinc Granular
ShelfLife 60 months