missing translation for 'onlineSavingsMsg'
Learn More

Zinc Dimethyldithiocarbamate 97.0+%, TCI America™

Numéro de catalogue. D071725G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
D071725G 25 g
D0717500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. D071725G Fournisseur TCI America Code fournisseur D071725G
Il en reste null

Identifiants chimiques

CAS 137-30-4
Molecular Formula C6H12N2S4Zn
Molecular Weight (g/mol) 305.796
MDL Number MFCD00064797
InChI Key DUBNHZYBDBBJHD-UHFFFAOYSA-L
Synonym ziram, zinc dimethyldithiocarbamate, carbazinc, fuclasin, methazate, zarlate, aazira, cymate, tsimat, zinc bis dimethyldithiocarbamate
PubChem CID 8722
ChEBI CHEBI:79736
IUPAC Name zinc;N,N-dimethylcarbamodithioate
SMILES CN(C)C(=S)[S-].CN(C)C(=S)[S-].[Zn+2]

Spécifications

Melting Point 257°C
Color White
Quantity 25 g
UN Number 2811
Formula Weight 305.80
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Zinc Dimethyldithiocarbamate
EINECSNumber (2)-1833&(9)-0607
RTECSNumber ZH0525000
TSCA Yes