missing translation for 'onlineSavingsMsg'
Learn More

Zinc chromate

Catalog No. AAA1817830
Change view
Click to view available options
Quantity:
250 g
1000 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1817830 250 g
AAA181780B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1817830 Supplier Thermo Scientific Chemicals Supplier No. A1817830

CAS: 13530-65-9 | CrO4Zn | 181.37 g/mol

Zinc chromate is used in pigments, rubber, linoleum manufacturing, metal surface treatment, electroplating intermediates, research and development. It finds its application as an useful metal for preventing corrosion when applied to other metals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Zinc chromate is used in pigments, rubber, linoleum manufacturing, metal surface treatment, electroplating intermediates, research and development. It finds its application as an useful metal for preventing corrosion when applied to other metals.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 13530-65-9
Molecular Formula CrO4Zn
Molecular Weight (g/mol) 181.37
MDL Number MFCD00036281
InChI Key NDKWCCLKSWNDBG-UHFFFAOYSA-N
Synonym zinc chromate, zinc chrome, zinc chromate c, zinc chromate o, zinc chromate t, zinc chromate z, zinc chromate am, zincro zto, basic zinc chromate, zinc chrome yellow
PubChem CID 26089
IUPAC Name zinc;dioxido(dioxo)chromium
SMILES [Zn++].[O-][Cr]([O-])(=O)=O

Specifications

Density 3.4
Quantity 250 g
UN Number UN3077
Solubility Information Insoluble in water.
Formula Weight 181.37
Chemical Name or Material Zinc chromate
Hazard Category H302-H317-H350i
Hazard Statement GHS H Statement
H350-H302-H317
May cause cancer.
Harmful if swallowed.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P261-P264b-P270-P272-P280g-P281-P301+P312-P302+P352-P308+P313-P330-P333+P313-P363-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 236-878-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only