missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc Chloride 98.0+%, TCI America™
Specifications
Specifications
| Boiling Point | 732°C |
| Melting Point | 290°C |
| Color | White |
| Physical Form | Crystalline Powder |
| Quantity | 25 g |
| UN Number | 2331 |
| Formula Weight | 136.28 |
| Percent Purity | ≥98.0% (T) |
| Chemical Name or Material | Zinc Chloride |
Safety and Handling
| RTECSNumber | ZH1400000 |
| TSCA | Yes |