missing translation for 'onlineSavingsMsg'
Learn More

Zinc chloride hydrate, 99.99% (metals basis)

Catalog No. AA4124730
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4124730 250 g
AA4124709 10 g
AA4124718 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4124730 Supplier Thermo Scientific Chemicals Supplier No. 04124730

CAS: 29604-34-0 | Cl2Zn | 136.28 g/mol

Zinc(II) chloride hydrate is used as a soldering flux and metal etchant. It is also employed in preserving anatomical specimens, wood preservatives, deodorant, disinfecting and embalming materials. It is employed as a mordant in printing and dyeing materials, and in the vulcanizing processes of fiber and rubber. It forms a complex with Ruhemann’s purple, which obtained by reacting ninhydrin with amino acids and amines is used in fingerprint detection.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 29604-34-0
Molecular Formula Cl2Zn
Molecular Weight (g/mol) 136.28
MDL Number MFCD00011295
InChI Key JIAARYAFYJHUJI-UHFFFAOYSA-L
Synonym zinc chloride, zinc dichloride, zinc chloride zncl2, zinc butter, zinc chloride fume, zinc ii chloride, zinkchloride, zintrace, zinc chloride, anhydrous, zine dichloride
PubChem CID 5727
ChEBI CHEBI:49976
IUPAC Name dichlorozinc
SMILES Cl[Zn]Cl

Specifications

Physical Form Crystalline Aggregates
Quantity 250 g
Assay Percent Range 99.99% (metals basis)
Solubility Information Soluble in water.
Formula Weight 136.29 (Anhydrous)
Chemical Name or Material Zinc chloride hydrate
Hazard Category H302-H314-H335
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
EINECSNumber 231-592-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only