missing translation for 'onlineSavingsMsg'
Learn More

Zinc Chloride, Granular, ACS, 97%, Spectrum™ Chemical

$215.59 - $12870.53

Chemical Identifiers

CAS 7646-85-7
View More Specs

Products 6
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18614728
View Documents
Spectrum Chemical Mfg Cor
Z105525GM
25 g Amber Glass Bottle
Each for $215.59
Only null left
 
18614726
View Documents
Spectrum Chemical Mfg Cor
Z1055125GM
125 g Amber Glass Bottle
Each for $274.06
Only null left
 
18614730
View Documents
Spectrum Chemical Mfg Cor
Z1055500GM
500 g Amber Glass Bottle
Each for $851.47
Only null left
 
18614731
View Documents
Spectrum Chemical Mfg Cor
Z105525KG
2.5 kg Amber Glass Bottle
Each for $2,760.91
Only null left
 
18614727
View Documents
Spectrum Chemical Mfg Cor
Z105512KG
12 kg Poly Pail
Each for $7,877.94
Only null left
 
18614729
View Documents
Spectrum Chemical Mfg Cor
Z105545KGBL
45 kg Fiber Drum N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

7646-85-7

Specifications

7646-85-7
25 g
ACS
1
97%
Amber Glass Bottle
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.