missing translation for 'onlineSavingsMsg'
Learn More

Zinc Chloride, Granular, ACS, 97%, Spectrum™ Chemical

Catalog No. 18614728
Change view
Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
12 kg
45 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18614728 25 g Amber Glass Bottle
18614726 125 g Amber Glass Bottle
18614730 500 g Amber Glass Bottle
18614731 2.5 kg Amber Glass Bottle
18614727 12 kg Poly Pail
18614729 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18614728 Supplier Spectrum Chemical Mfg Cor Supplier No. Z105525GM
Only null left

Chemical Identifiers

CAS 7646-85-7

Specifications

CAS Max % 1
Quantity 25 g
Percent Purity 97%
Grade ACS
Packaging Amber Glass Bottle

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.