missing translation for 'onlineSavingsMsg'
Learn More

Zinc Chloride, Granular, ACS, 97%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor Z105512KG

 View more versions of this product

Catalog No. 18614727


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

7646-85-7
12 kg
ACS
1
97%
Poly Pail

Chemical Identifiers

7646-85-7
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.