missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chlorure de zinc (Cryst./ACS certifié), Fisher Chemical™
Spécifications
Spécifications
| Point d’ébullition | 732 °C |
| Nom ou substance chimique | Zinc Chloride |
| Point de fusion | 293 °C |
| Couleur | Blanc |
| pH | 5 |
| Forme physique | Solide |
| Quantité | 100 g |
| Fourchette de pourcentages d’essai | ≥97 % |
| Numéro UN | 2331 |
| Poids de formule | 136.3 |
| Afficher plus de résultats |
Sécurité et manipulation
| Hazard Statement | DANGER! |
| Precautionary Statement | Emergency Overview Causes burns by all exposure routes. Very toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Hygroscopic. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required. NFPA Health:3 Flammability:1 Instability:1 |
| DOTInformation | DOT Class 8, : Corrosive |