missing translation for 'onlineSavingsMsg'
Learn More

Zinc Bromide 98.0+%, TCI America™

Catalog No. Z0013300G
Change view
Click to view available options
Quantity:
25 g
300 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
Z0013300G 300 g
Z001325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. Z0013300G Supplier TCI America Supplier No. Z0013300G

Chemical Identifiers

CAS 7699-45-8
Molecular Formula Br2Zn
Molecular Weight (g/mol) 225.188
MDL Number MFCD00011294
InChI Key VNDYJBBGRKZCSX-UHFFFAOYSA-L
Synonym zincbromide, zinc ii bromide, acmc-209p7h, zinc 2+ ion dibromide, ksc378m6n, zinc bromide hydrate, puratronic™, zinc bromide, anhydrous 100g
PubChem CID 10421208
IUPAC Name zinc;dibromide
SMILES [Zn+2].[Br-].[Br-]

Specifications

Boiling Point 697°C
Melting Point 394°C
Color White
Physical Form Crystalline Powder
Quantity 300 g
UN Number 3260
Formula Weight 225.19
Percent Purity ≥98.0% (T)
Chemical Name or Material Zinc Bromide
EINECSNumber (1)-0118
RTECSNumber ZH1150000
TSCA Yes