missing translation for 'onlineSavingsMsg'
Learn More

Zinc, ACS Grade, LabChem™

Catalog No. LC270607
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC270607 100 g Poly Bottle
LC270601 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. LC270607 Supplier LabChem Supplier No. LC270607

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7440-66-6
Molecular Formula Zn
Molecular Weight (g/mol) 65.38
MDL Number MFCD00011291
InChI Key HCHKCACWOHOZIP-UHFFFAOYSA-N
Synonym dust, cinc, powder, merrillite, rheinzink, jasad, blue powder, granular, zinc, elemental, emanay dust
PubChem CID 23994
ChEBI CHEBI:30185
IUPAC Name zinc
SMILES [Zn]

Specifications

Color Silver
Melting Point 419°C
Boiling Point 907°C
CAS Max % 100
Quantity 100 g
Identification Passes Test
Mesh Size 20 Mesh
Packaging Poly Bottle
Form Granular
Linear Formula Zn
Formula Weight 65.38
Grade ACS
Chemical Name or Material Zinc
Show More Show Less
Hazard Statement GHS H Statement
Product is not hazardous.
Precautionary Statement GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
EINECSNumber 231-175-3
Recommended Storage Room Temperature