missing translation for 'onlineSavingsMsg'
Learn More

Zinc acetate, anhydrous, 99.98% (metals basis)

Catalog No. AA3579218
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3579218 50 g
AA3579209 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3579218 Supplier Thermo Scientific Chemicals Supplier No. 03579218

CAS: 557-34-6 | C4H6O4Zn | 183.468 g/mol

Zinc acetate anhydrous is used in the synthesis of layered Zn-arylphosphonates with potential application in sorption, ion exchange or catalysis. It is utilized in the ultrasonic preparation of zinc sulfide nanoparticles coated on silica particles. It is administered orally or parenterally as a nutritional supplement. It finds an application in the field of industries such as wood preservation, manufacturing other zinc salts, polymers, manufacture of ethylene acetate, as a dye mordant, and analytical reagent. It also acts as a plating inhibitor on primary water piping.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 557-34-6
Molecular Formula C4H6O4Zn
Molecular Weight (g/mol) 183.468
MDL Number MFCD00012454
InChI Key DJWUNCQRNNEAKC-UHFFFAOYSA-L
Synonym zinc acetate, zinc diacetate, zinc ii acetate, acetic acid, zinc salt, dicarbomethoxyzinc, acetic acid, zinc ii salt, zinc di acetate, galzin, zinc acetate anhydrous, siltex cl 4
PubChem CID 11192
ChEBI CHEBI:62984
IUPAC Name zinc;diacetate
SMILES CC(=O)[O-].CC(=O)[O-].[Zn+2]

Specifications

Melting Point 200°C (decomposition)
Physical Form Granules
Quantity 50 g
Assay Percent Range 99.98% (metals basis)
Solubility Information Soluble in water,alcohol,dilute mineral acids and alkalies.
Formula Weight 183.47
Sensitivity Hygroscopic
Density 1.84 g/mL
Chemical Name or Material Zinc acetate
Hazard Category H302-H318
Precautionary Statement P264b-P270-P280-P301+P312-P305+P351+P338-P310-P330-P501c
DOTInformation Hazard Class: 9; Packaging Group: III
EINECSNumber 209-170-2
RTECSNumber AK1500000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only