missing translation for 'onlineSavingsMsg'
Learn More

Zinc, AAS standard solution, Specpure™ Zn 1000μg/mL

Catalog No. AA88118AE
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA88118AE 100 mL
AA88118AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA88118AE Supplier Thermo Scientific Chemicals Supplier No. 088118AE

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

MDL Number MFCD00011291

Specifications

Physical Form Liquid
Concentration Zn in 5% HNO3
UN Number UN3264
Quantity 100 mL
Grade Specpure
Chemical Name or Material Zinc, AAS standard solution, Zn 1000μg/mL
Hazard Category H290-H314-H335
Hazard Statement GHS H Statement
H314
Causes severe skin burns and eye damage.
Precautionary Statement P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, INORGANIC, N.O.S.
EINECSNumber 231-714-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only