Learn More
Zinc AA Standard, Certified, 1000ppm ±10ppm (1mL = 1mg Zn), LabChem™
$74.64 - $158.21
Chemical Identifiers
| CAS | 10196-18-6 |
|---|---|
| Molecular Formula | H12N2O12Zn |
| Molecular Weight (g/mol) | 297.48 |
| MDL Number | MFCD00149889 |
| InChI Key | JGPSMWXKRPZZRG-UHFFFAOYSA-N |
| PubChem CID | 15865313 |
| IUPAC Name | zinc(2+) hexahydrate dinitrate |
| SMILES | O.O.O.O.O.O.[Zn++].[O-][N+]([O-])=O.[O-][N+]([O-])=O |
Chemical Identifiers
| 10196-18-6 | |
| 297.48 | |
| JGPSMWXKRPZZRG-UHFFFAOYSA-N | |
| zinc(2+) hexahydrate dinitrate |
| H12N2O12Zn | |
| MFCD00149889 | |
| 15865313 | |
| O.O.O.O.O.O.[Zn++].[O-][N+]([O-])=O.[O-][N+]([O-])=O |
Specifications
| Poly Bottle | |
| 93.54,6,0.46 | |
| 1.05g/mL | |
| H12N2O12Zn | |
| Zn(NO3)2·6H2O | |
| UN3264 | |
| Soluble in water | |
| O.O.O.O.O.O.[Zn++].[O-][N+]([O-])=O.[O-][N+]([O-])=O | |
| 297.48 | |
| 15865313 | |
| Certified |
| 10196-18-6 , 7697-37-2 , 7732-18-5 | |
| Passes Test | |
| Liquid | |
| 1000 ppm ±10 ppm (1mL = 1mg Zn) | |
| MFCD00149889 | |
| Colorless | |
| JGPSMWXKRPZZRG-UHFFFAOYSA-N | |
| zinc(2+) hexahydrate dinitrate | |
| 125 mL | |
| 297.48 | |
| Zinc AA Standard, 1000ppm (1mL = 1mg Zn) |
Safety and Handling
GHS H Statement
May be corrosive to metals.
Causes severe skin burns and eye damage.
Fatal if inhaled.
GHS P Statement
Keep only in original container.
Do not breathe mist, vapors, spray.
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Absorb spillage to prevent material damage.
Store locked up.
Store in corrosive resistant container with a resistant inner liner.
Dispose of contents/container to comply with local, state and federal regulations.
Danger
Recommended Storage : Room Temperature