missing translation for 'onlineSavingsMsg'
Learn More

Zinc AA Standard, Certified, 1000ppm ±10ppm (1mL = 1mg Zn), LabChem™

Catalog No. LC271507
Change view
Click to view available options
Quantity:
125 mL
500 mL
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC271507 125 mL Poly Bottle
LC271501 500 mL Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. LC271507 Supplier LabChem Supplier No. LC271507

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 10196-18-6 , 7697-37-2 , 7732-18-5
Molecular Formula H12N2O12Zn
Molecular Weight (g/mol) 297.48
MDL Number MFCD00149889
InChI Key JGPSMWXKRPZZRG-UHFFFAOYSA-N
PubChem CID 15865313
IUPAC Name zinc(2+) hexahydrate dinitrate
SMILES O.O.O.O.O.O.[Zn++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Packaging Poly Bottle
CAS Max % 93.54, 6, 0.46
Identification Passes Test
Density 1.05g/mL
Physical Form Liquid
Concentration 1000 ppm ±10 ppm (1mL = 1mg Zn)
Linear Formula Zn(NO3)2·6H2O
UN Number UN3264
Color Colorless
Solubility Soluble in water
Quantity 125 mL
Formula Weight 297.48
Grade Certified
Chemical Name or Material Zinc AA Standard, 1000ppm (1mL = 1mg Zn)
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
May be corrosive to metals.
Causes severe skin burns and eye damage.
Fatal if inhaled.
Precautionary Statement GHS P Statement
Keep only in original container.
Do not breathe mist, vapors, spray.
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Absorb spillage to prevent material damage.
Store locked up.
Store in corrosive resistant container with a resistant inner liner.
Dispose of contents/container to comply with local, state and federal regulations.
Recommended Storage Room Temperature