Learn More
Restek™ Zero-Dead-Volume Valco Internal Reducing Union
Connect two sizes of tubing, using zero-dead-volume fittings on each end.
Supplier:  Restek™ RK 20151
Encompass
 Afficher plus de versions de ce produit
Numéro de catalogue. 06710519
Il en reste null
Description
Description
  • For 0.125 to 0.0625 in. O.D. tubing
  • Includes stainless steel ferrules
Spécifications
Identifiants chimiques
Union
HPLC
Spécifications
Union
0.25 mm
HPLC
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS