Learn More
MilliporeSigma™ Supelco™ Zenix™ SEC-300 Gel Filtration HPLC Column, 3 μm Particle Size, Stainless Steel
Like their 5 micron siblings, 3 micron Zenix™ SEC columns feature a hydrophilic surface layer to prevent the sample from interacting with the underlying silica.
$2638.58 - $3602.82
Specifications
Particle Size 3 μm
pH 2.0 to 8.5
Pore Size 300 Å
Stationary Phase Hydrophilic Polymer
Product Line Zenix
View More Specs
Products 9
Catalog Number Mfr. No. Length (Metric) Diameter (Metric) Price Quantity  
Catalog Number Mfr. No. Length (Metric) Diameter (Metric) Price Quantity  
111005961
View Documents
MilliporeSigma™ Supelco™
Z777022
30 cm 1 mm
Each for $3,602.82
Only null left
111005962
View Documents
MilliporeSigma™ Supelco™
Z777024
30 cm 2.1 mm
Each for $3,602.82
Only null left
111006253
View Documents
MilliporeSigma™ Supelco™
Z777026
15 cm 4.6 mm
Each for $2,795.62
Only null left
111006210
View Documents
MilliporeSigma™ Supelco™
Z777027
25 cm 4.6 mm
Each for $3,002.43
Only null left
111006062
View Documents
MilliporeSigma™ Supelco™
Z777028
30 cm 4.6 mm
Each for $3,175.72
Only null left
111006254
View Documents
MilliporeSigma™ Supelco™
Z777030
15 cm 7.8 mm
Each for $2,638.58
Only null left
111006209
View Documents
MilliporeSigma™ Supelco™
Z777031
20 cm 7.8 mm
Each for $2,990.08
Only null left
111006208
View Documents
MilliporeSigma™ Supelco™
Z777032
25 cm 7.8 mm
Each for $3,027.18
Only null left
111005963
View Documents
MilliporeSigma™ Supelco™
Z777033
30 cm 7.8 mm
Each for $3,602.82
Only null left
Description
Description
Select 100, 300 or 500 Å pores based on the size of your sample molecules, whether you are analyzing peptides, proteins, mAbs, ADCs, fusion proteins, etc. in quality control or research.
Specifications
Specifications
3 μm
300 Å
Zenix
Size Exclusion
SEC HPLC
2.0 to 8.5
Hydrophilic Polymer
Hydrophilic Polymer Phase/Spherical Silica
Stainless Steel
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'
Zenix is a registered trademark of Sepax Technologies