missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Zenix™ SEC-300 Gel Filtration HPLC Column, 3 μm Particle Size, Stainless Steel

Catalog No. 111005961
Change view
Click to view available options
Length (Metric):
15 cm
20 cm
25 cm
30 cm
Diameter (Metric):
1 mm
2.1 mm
4.6 mm
7.8 mm
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Diameter (Metric)
111005961 30 cm 1 mm
111005962 30 cm 2.1 mm
111006253 15 cm 4.6 mm
111006210 25 cm 4.6 mm
111006062 30 cm 4.6 mm
111006254 15 cm 7.8 mm
111006209 20 cm 7.8 mm
111006208 25 cm 7.8 mm
111005963 30 cm 7.8 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 111005961 Supplier MilliporeSigma™ Supelco™ Supplier No. Z777022

Like their 5 micron siblings, 3 micron Zenix™ SEC columns feature a hydrophilic surface layer to prevent the sample from interacting with the underlying silica.

Select 100, 300 or 500 Å pores based on the size of your sample molecules, whether you are analyzing peptides, proteins, mAbs, ADCs, fusion proteins, etc. in quality control or research.

Specifications

Diameter (Metric) 1 mm
Length (Metric) 30 cm
Particle Size 3 μm
pH 2.0 to 8.5
Pore Size 300 Å
Stationary Phase Hydrophilic Polymer
Product Line Zenix
Matrix Hydrophilic Polymer Phase/Spherical Silica
Phase Size Exclusion
Material Stainless Steel
Technique SEC HPLC
Show More Show Less

Zenix is a registered trademark of Sepax Technologies