missing translation for 'onlineSavingsMsg'
Learn More

(Z)-2-(2-Amino-4-thiazolyl)-2-(methoxyimino)acetic Acid 98.0+%, TCI America™

Catalog No. A24785G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A24785G 5 g
A247825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A24785G Supplier TCI America Supplier No. A24785G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 65872-41-5
Molecular Formula C6H6N3O3S
Molecular Weight (g/mol) 200.19
MDL Number MFCD00071528,MFCD00010416
InChI Key NLARCUDOUOQRPB-WTKPLQERSA-M
Synonym 2-amino-4-thiazolyl-methoxyimino acetic acid, 2-2-aminothiazole-4-yl-2-methoxyiminoacetic acid, 2-2-aminothiazol-4-yl-2-methoxyimino acetic acid, z-2-2-aminothiazol-4-yl-2-methoxyimino acetic acid, pubchem19489, z-2-aminothiazole-4-yl-2-methoxyiminoacetic acid, syn-2-2-aminothiazol-4-yl-2-methoxyiminoacetic acid, 2z-2-amino-1,3-thiazol-4-yl methoxyimino acetic acid, 2z-2-2-aminothiazol-4-yl-2-methoxyimino-acetic acid, nlarcudouoqrpb-wtkplqersa-n
PubChem CID 5582975
IUPAC Name (2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-(methoxyimino)acetate
SMILES CO\N=C(/C([O-])=O)C1=CSC(N)=N1

Specifications

Melting Point 204°C
Color Yellow
Quantity 5 g
Formula Weight 201.20
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material (Z)-2-(2-Amino-4-thiazolyl)-2-(methoxyimino)acetic Acid