missing translation for 'onlineSavingsMsg'
Learn More

Yttrium ICP Standard, 1000 ppm Y in 3% HNO3, Ricca Chemical

Catalog No. PY1KN500
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Natural LDPE Bottle
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
PY1KN500 500 mL Natural Poly Bottle
PY1KN100 100 mL Natural LDPE Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PY1KN500 Supplier Ricca Chemical Company Supplier No. PY1KN500

Chemical Identifiers

CAS 1314-36-9 , 7697-37-2 , 7732-18-5
Molecular Formula O3Y2
Molecular Weight (g/mol) 225.81
MDL Number MFCD00011473
InChI Key RUDFQVOCFDJEEF-UHFFFAOYSA-N
IUPAC Name diyttrium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Y+3].[Y+3]

Specifications

Packaging Natural Poly Bottle
CAS Min % 93.0, 4.8, 0.12
CAS Max % 97.0, 4.99, 0.12
Physical Form Liquid
Color Colorless
pH < 2
Melting Point Approximately 0°C
Type Standard
Quantity 500 mL
Concentration or Composition (by Analyte or Components) 1mL = 1mg Y (1, 000ppm Y)Y2O3 in 3% HNO3
Chemical Name or Material Yttrium ICP Standard
Show More Show Less
ShelfLife 18 months