missing translation for 'onlineSavingsMsg'
Learn More

Yttrium ICP-MS Standard, 1000 ppm Y in 3% HNO3, Ricca Chemical

Catalog No. MSY1KN100
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MSY1KN100 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MSY1KN100 Supplier Ricca Chemical Company Supplier No. MSY1KN100

Chemical Identifiers

CAS 1314-36-9 , 7697-37-2 , 7732-18-5
Molecular Formula O3Y2
Molecular Weight (g/mol) 225.81
MDL Number MFCD00011473
InChI Key RUDFQVOCFDJEEF-UHFFFAOYSA-N
IUPAC Name diyttrium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Y+3].[Y+3]

Specifications

Packaging Natural LDPE Bottle
Concentration or Composition Notes 1,000ppm Y
CAS Min % 93.0, 4.8, 0.12
CAS Max % 97.0, 4.99, 0.12
Equivalence 1 mL = 1 mg Y
Physical Form Liquid
Color Colorless
Quantity 100 mL
Chemical Name or Material Yttrium ICP-MS Standard