missing translation for 'onlineSavingsMsg'
Learn More

Yttrium AA Standard, 1000 ppm Y in 3% HNO3, Ricca Chemical

Catalog No. AY1KN100
Change view
Click to view available options
Quantity:
100 mL
Packaging:
Natural LDPE Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AY1KN100 100 mL Natural LDPE Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AY1KN100 Supplier Ricca Chemical Company Supplier No. AY1KN100

Chemical Identifiers

CAS 1314-36-9 , 7697-37-2 , 7732-18-5
Molecular Formula O3Y2
Molecular Weight (g/mol) 225.81
MDL Number MFCD00011473
InChI Key RUDFQVOCFDJEEF-UHFFFAOYSA-N
IUPAC Name diyttrium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Y+3].[Y+3]

Specifications

Packaging Natural LDPE Bottle
CAS Min % 93.0, 4.8, 0.12
CAS Max % 97.0, 4.99, 0.12
Physical Form Liquid
Color Colorless
pH < 2
Melting Point Approximately 0°C
Type Standard
Quantity 100 mL
Concentration or Composition (by Analyte or Components) 1mL = 1mg Y (1, 000ppm Y)Y2O3 in 3% HNO3
Chemical Name or Material Yttrium AA Standard
Show More Show Less
ShelfLife 24 months