missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ytterbium ICP Standard, 1000 ppm Yb in 3% HNO3, Ricca Chemical
Specifications
Specifications
| Packaging | Natural LDPE Bottle |
| Concentration or Composition Notes | 1,000ppm Yb |
| CAS Min % | 94.0, 4.36, 0.11 |
| CAS Max % | 97.0, 4.54, 0.11 |
| Equivalence | 1 mL = 1 mg Yb |
| Chemical Name or Material | Ytterbium ICP Standard |