CAS RN 1314-37-0
IUPAC Name:
diytterbium(3+) trioxidandiide
Synonyms:
ytterbium(III) oxide
Molecular Weight (g/mol):
394.09
Molecular Formula:
O3Yb2
InChi Key:
UZLYXNNZYFBAQO-UHFFFAOYSA-N
SMILES:
[O--].[O--].[O--].[Yb+3].[Yb+3]
1
–
5
of
5
results
1
Ytterbium(III) oxide, REacton™, 99.9% (REO)
CAS: 1314-37-0 Molecular Formula: O3Yb2 Molecular Weight (g/mol): 394.09 MDL Number: MFCD00011290 InChI Key: UZLYXNNZYFBAQO-UHFFFAOYSA-N Synonym: Ytterbia IUPAC Name: diytterbium(3+) trioxidandiide SMILES: [O--].[O--].[O--].[Yb+3].[Yb+3]
2
Ytterbium(III) oxide, REacton™, 99.99% (REO)
CAS: 1314-37-0 Molecular Formula: O3Yb2 Molecular Weight (g/mol): 394.09 MDL Number: MFCD00011290 InChI Key: UZLYXNNZYFBAQO-UHFFFAOYSA-N IUPAC Name: diytterbium(3+) trioxidandiide SMILES: [O--].[O--].[O--].[Yb+3].[Yb+3]
4
5
Ytterbium(III) Oxide 98.0+%, TCI America™
CAS: 1314-37-0 Molecular Formula: O3Yb2 Molecular Weight (g/mol): 394.09 MDL Number: MFCD00011290 InChI Key: UZLYXNNZYFBAQO-UHFFFAOYSA-N IUPAC Name: diytterbium(3+) trioxidandiide SMILES: [O--].[O--].[O--].[Yb+3].[Yb+3]