missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ YTHDC1 Polyclonal Antibody
GREENER_CHOICE

Numéro de catalogue. PIPA557670
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
PIPA557670 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. PIPA557670 Fournisseur Invitrogen™ Code fournisseur PA557670
Il en reste null

Rabbit Polyclonal Antibody

Immunogen sequence: LVSKPLSSSV SNNKRIVSTK GKSATEYKNE EYQRSERNKR LDADRKIRLS SSASREPYKN QPEKTCVRKR DPERRAKSPT PDGSERIGLE VD Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 92%.

May be part of a signal transduction pathway that influences splice site selection. Target protein binds nuclear transcriptosomal complex scaffold attachment factor B (SAF-B) and Sam68; may play a role in splice site selection and alternative splicing.

Spécifications

Antigen YTHDC1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene YTHDC1
Gene Accession No. Q96MU7
Gene Alias A730098D12Rik; C80342; KIAA1966; mKIAA1966; putative splicing factor YT521; RA301-binding protein; Splicing factor YT521; splicing factor YT521-B; YT521; YT521-B; YTH domain containing 1; YTH domain-containing protein 1; YTHDC1
Gene Symbols YTHDC1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human YTHDC1. Recombinant protein control fragment (Product #RP-96608).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 91746
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Afficher plus de résultats Afficher moins de résultats