missing translation for 'onlineSavingsMsg'
Learn More
Learn More
YSI™ pH/ORP Dual Probes for Pro Series Portable Meters
For use with Professional Plus Multiparameter Handheld Instrument.
Supplier: YSI™ 605103
Description
- Easy user-replaceble sensor
- Measure pH and ORP or redox quickly and easily
- Requires cable/probe assembly
Specifications
| pH/ORP probe | |
| Professional Plus Multiparameter Handheld Instrument | |
| pH/ORP dual probe |