Learn More
YSI™ MultiLab Line BOD/pH Laboratory Kits
Catalog No. 09324568
Change view
Click to view available options
Channels:
2
Catalog No. Channels
09324568 2
1 options
Catalog No. 09324568 Supplier YSI™ Supplier No. 626573
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Kit for BOD/pH calculation
BOD/pH Lab Kit Includes
  • 4010-2 Two Channel benchtop instrument
  • Double Junction pH electrode
  • Optical BOD probe
Advanced BOD/pH Lab Kit Includes
  • 4010-2 Two Channel benchtop instrument
  • Double Junction pH electrode
  • Optical BOD probe
  • BOD Analyst Pro desktop software
Specifications
Channels 2
Includes 4010-2 Two channel benchtop instrument, optical BOD probe and double junction pH electrode