missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Youngstown Barrel & Drum Tight Head Steel Drum
Youngstown Tight Head Steel Pail is perfect for the transporting of hazardous solids or liquids.
Supplier: Youngstown Barrel & Drum YBD2322
Description
With cold-rolled carbon-steel construction, steel drums are qualified for safe transport of noncorrosive hazardous liquids or solids.
- Salvage drum designed to overpack and ship damaged and/or leaking drums
- Also qualify for direct containment of hazardous solids
Identifiants chimiques
| Black | |
| 50 | |
| Tight | |
| Ea. |
Spécifications
| UN 1A1/Y1.8/100-DOT/UN | |
| 24 Gauge | |
| Black | |
| Steel | |
| Tight | |
| 13.5 in. | |
| 10.75 to 11.5 in. |
| Pail | |
| 18.92 L | |
| For transport of hazardous solids or liquids, Designed to overpack and ship damaged and/or leaking drums | |
| 50 | |
| Ea. | |
| 34.29 cm | |
| 27.3 to 29.21 mm |