missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Youngstown Barrel & Drum Tight Head Steel Drum
Youngstown Tight Head Steel Pail is perfect for the transporting of hazardous solids or liquids.
Supplier: Youngstown Barrel & Drum YBD345
Description
With cold-rolled carbon-steel construction, steel drums are qualified for safe transport of noncorrosive hazardous liquids or solids.
- Salvage drum designed to overpack and ship damaged and/or leaking drums
- Also qualify for direct containment of hazardous solids
Identifiants chimiques
| Black | |
| 50 | |
| Tight | |
| Ea. |
Spécifications
| UN 1A1/X1.2/300-DOT/UN | |
| Bung | |
| 208.19 L | |
| For transport of hazardous solids or liquids, Designed to overpack and ship damaged and/or leaking drums | |
| 50 | |
| Ea. | |
| 86.36 cm | |
| 57.15 mm | |
| 15.87 Kg |
| Unlined Drum | |
| 20/18 Gauge | |
| Black | |
| Carbon Steel | |
| Tight | |
| 34 in. | |
| 22.5 in. | |
| 35 lb. |